Learn More
Description
- Available in 12 colors
- Etching on top surface for writing purpose
- Fits on top of Cryogenic vial caps as well as sample tube caps T500, T501 and T502
- Drain hole on top to prevent LN2 buildup
Specifications
Specifications
| Color | Violet |
| Material | Polypropylene |
| For Use With (Application) | Cap of the Cryovial™ tubes for color identification |
| Packaging | Bag |
| Quantity | 500 Pk. |
| Product Type | Cap Insert |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.