Learn More
Simport™ Scientific SecurTainer™ I Tamper Evident Specimen Container
Catalog No. 22038630 Shop All Simport Scientific Products
Change view
Click to view available options
Capacity (Metric):
20 mL
40 mL
60 mL
90 mL
120 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
22-038-630 20 mL
22-038-631 40 mL
22-038-633 90 mL
22-038-634 120 mL
22-038-632 60 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 22-038-630 Supplier Simport™ Scientific Supplier No. C57520MA
Add to Cart
Add to Cart
SecurTainer™ I Tamper Evident Specimen Container for collection, transport and storage of specimens.
  • Shatter resistant polypropylene containers
  • Latex free
  • Closure made of polyethylene
  • Incorporates unique tamper evident screw cap ensuring safe transport or storage situations
  • For transporting specimens
Specifications
Material Polypropylene
Sterility Non-sterile
Capacity (Metric) 20 mL
Closure Color Magenta
Closure Material Polyethylene
Closure Type Ribbed Tamper-evident Screw Cap
No. per Case 500/Cs.
No. per Pack 100 Pk.