Learn More
Simport™ Scientific SecurTainer™ I Tamper Evident Specimen Container
Shop All Simport Scientific ProductsDescription
- Shatter resistant polypropylene containers
- Latex free
- Closure made of polyethylene
- Incorporates unique tamper evident screw cap ensuring safe transport or storage situations
- For transporting specimens
Specifications
Specifications
| Material | Polypropylene |
| Sterility | Non-sterile |
| Capacity (Metric) | 20 mL |
| Closure Color | Magenta |
| Closure Material | Polyethylene |
| Closure Type | Ribbed Tamper-evident Screw Cap |
| No. per Case | 500/Cs. |
| No. per Pack | 100 Pk. |