Learn More
Simport™ Scientific SLIMSETTE™ M510 Biopsy Processing/Embedding Cassettes
Catalog No. 23045165 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Pink
Catalog No. Color
23-045-165 Pink
1 options
Catalog No. 23-045-165 Supplier Simport™ Scientific Supplier No. M5103
Add to Cart
Add to Cart
SLIMSETTE™ M510 Biopsy Processing/Embedding Cassettes are a new generation of cassettes that are more compact and easier to use.
  • Specially designed to hold biopsy specimens during the embedding process
  • Made of acetal polymer
  • Unique recessed cover
  • Cassettes are thinner, allowing more to be stacked in automatic labeling machines
  • One-piece integral lid
  • Large labeling area for easy identification
  • Anterior writing area at a 45° angle
Specifications
Quantity 3 x 500/Cs.
Writing Face Angle 45°
Closure Type Integral Lid
Color Pink
Material Acetal Polymer
Product Type Biopsy Cassette