Learn More
Simport™ Scientific Swingsette™ Tissue Cassettes In Quickload™ Sleeves
Catalog No. 22038640 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Aqua
Gray
Green
Lilac
Orange
Peach
Tan
White
Yellow
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-038-640 White
22-038-643 Peach
22-038-644 Tan
22-038-645 Gray
22-038-647 Aqua
22-038-646 Orange
22-038-263 Yellow
22-038-264 Lilac
22-038-266 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 22-038-640 Supplier Simport™ Scientific Supplier No. M5172SL
Add to Cart
Add to Cart
Suitable for hoppers accepting plastic sleeves, these Swingsette™ Tissue Cassettes In Quickload™ Sleeves will load in cassette labeling instruments in one simple operation. Just load the 75-cassette QuickLoad™ sleeve in the hopper and you are ready for printing.
  • Made of acetal
  • Sleeved cassettes for use with Thermo Fisher printers
  • Anterior writing area at a 45° angle
  • Large tab for convenient and easy opening
  • Covers packaged separately
  • Easy-to-assemble disposable cover with special hinge
Specifications
Color White
Product Type Tissue Cassette