Learn More
Simport™ Scientific Swingsette™ Tissue Processing/Embedding Cassettes with Hinged Covers
Catalog No. 22038661 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Yellow
Catalog No. Color
22-038-661 Yellow
1 options
Catalog No. 22-038-661 Supplier Simport™ Scientific Supplier No. M5155
Add to Cart
Add to Cart
Disposable plastic Swingsette™ Tissue Processing/Embedding Cassettes hold tissue specimens during processing, embedding and storage.
  • Molded from a special high-density polymer, the cassettes keep specimens safely submerged in liquid and are totally resistant to the chemical action of histological solvents
  • Special hinge holds base and cover together, allowing cassettes to be opened and closed repeatedly
  • Cover can be removed and re-inserted easily without danger of specimen loss
  • Efficient flow-through slots maximize fluid exchange and ensure proper drainage
Specifications
Closure Included Yes
Color Yellow
Material High Density Polymer
Product Type Tissue Cassette