Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simulaids™ Adult Airway Management Trainer Torso
SDPSCI_ED

Catalog No. 19819206 Shop All Simulaids, Inc. Products
Change view
Click to view available options
For Use With (Application):
Advanced Airway Management Procedures; CPR
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
19-819-206 Advanced Airway Management Procedures; CPR
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-819-206 Supplier Simulaids™ Supplier No. 101086
Add to Cart
Add to Cart

Includes

Oral and nasal airway for intubation, esophagus and inflatable lungs and stomach

Features anatomical landmarks including sternum, rib cage and substernal notch

  • Advanced life support manikin
  • Contains teeth, uvula, vocal cords, glottis, epiglottis, larynx, arytenoids cartilage, and trachea
  • Size: 30 x 17 x 10 in. (76.2 x 43 x 25.4cm)
  • wt.: 20 lb. (9kg)

Specifications

For Use With (Application) Advanced Airway Management Procedures; CPR
Includes Anatomical landmarks (sternum, rib cage, substernal notch); teeth, uvula, vocal cords, glottis, epiglottis, larynx, arytenoid cartilage, trachea, esophagus, and inflatable lungs and stomach
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.