Learn More
Simulaids™ Defibrillation/CPR Trainer
SDPSCI_ED
Catalog No. 19819207 Shop All Simulaids, Inc. Products
Click to view available options
Includes:
Manual, semiautomatic, automatic defibrillators
Catalog No. Includes
19-819-207 Manual, semiautomatic, automatic defibrillators
1 options
Catalog No. 19-819-207 Supplier Simulaids™ Supplier No. 101100
Add to Cart
Add to Cart
Request a Quote
Includes
10 mouth/nose pieces, 10 airway systems with lung, adapter numbers: 050, 051, 052, ALS chest post set, arrhythmia simulator, and soft carry bag.
For demonstration and practice of defibrillation and CPR training
  • Functions with manual, semiautomatic, and automatic defibrillators
  • The arrhythmia/pacing simulator
  • Capable of recreating 17 adult heart rhythms and 17 pediatric rhythms
  • Electronic capture can be simulate with external pacer
  • Also simulate cardioversion with manual, semiautomatic, or automatic defibrillation
Specifications
Product Type Defibrillation/CPR Training Mannequin with Carry Bag
For Use With (Application) Defibrillation and CPR Training
Includes Manual, semiautomatic, automatic defibrillators
WARNING: Cancer - www.P65Warnings.ca.gov