Learn More
Simulaids™ Economy Adult Sani-Manikin
SDPSCI_ED
Catalog No. 19500461 Shop All Simulaids, Inc. Products
Click to view available options
Includes:
Carry Bag
Without Carry Bag
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
19-500-461 Carry Bag
19-500-460 Without Carry Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-500-461 Supplier Simulaids™ Supplier No. 1002145
Designed with the instructor in mind — no cleaning, disinfecting or disassembly required
Lightweight, yet durable for extended training life
  • Realistic head tilt
  • Single-use airway/lung/face shield system
  • Simulated chest rise, palpable landmarks, and xiphoid process for reference in hand placement
  • Can also be used for AED training
  • Latex free
Order Info
Face Shield Lung System sold separately.
Specifications
Product Type Economy Adult Sani-Manikin
For Use With (Application) AED Training
Includes Carry Bag
Weight (English) 28 lb.
WARNING: Cancer - www.P65Warnings.ca.gov