Learn More
Simulaids™ IV Training Arm and Hand
SDPSCI_ED
Catalog No. 19819217 Shop All Simulaids, Inc. Products
Click to view available options
IV Training arm and hand; 18 x 6 x 5 in.
Catalog No.
19-819-217 IV Training arm and hand; 18 x 6 x 5 in.
1 options
Catalog No. 19-819-217 Supplier Simulaids™ Supplier No. 140146
Includes
Carry tray and blood
Designed for practice in giving IVs
  • Permits detailed exercises in venipuncture for getting expertise on realistic model before actually administering to patients
  • Features lifelike vein roll-over and allows repeated palpation of vein for location
  • Blood can be withdrawn or fluid injected
  • When needle penetrates into vein, characteristic pop can be felt
Specifications
Product Type I.V. Training Arm
Includes Lifelike vein roll-over, durable for repeated use, palpation of vein for location
WARNING: Cancer - www.P65Warnings.ca.gov