Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simulated Gastric Fluid (without Pepsin), Ricca Chemical
SDP

Catalog No. 710825 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
7108-25 10 L Cubitainer
7108-16 500 mL Natural Poly Bottle
7108-32 1 L Natural Poly Bottle
7108-5 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 7108-25 Supplier Ricca Chemical Company Supplier No. 71082.5
Add to Cart
Add to Cart

0.2% (w/v) Sodium Chloride in 0.7% (v/v) Hydrochloric Acid

Specifications

Chemical Name or Material Simulated Gastric Fluid (without Pepsin)
Color Colorless
CAS 7732-18-5, 7647-01-0, 7647-14-5
CAS Min % 97.0, 0.8, 0.19
CAS Max % 100.0, 0.83, 0.2
Percent Purity 0.2%
Grade Laboratory
Packaging Cubitainer
Solvent or Matrix 0.7% (v/v) Hydrochloric Acid
pH 1 to 1.4
Quantity 10 L
Physical Form Liquid
Concentration or Composition Notes 0.2% (w/v) Sodium Chloride in 0.7% (v/v) Hydrochloric Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.