Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simulated Intestinal Fluid (without Pancreatin), USP XXII Formulation, Ricca Chemical
SDP

Simulated Intestinal Fluid (without Pancreatin)

$189.70 - $1020.00

Specifications

CAS 7732-18-5 , 7778-77-0 , 1310-73-2
Grade Laboratory
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
710975-16
SDP
View Documents
Ricca Chemical Company
7109.7516
500 mL Natural Poly Bottle
Each for $189.70
Add to Cart
 
R710975020F
SDP
Accessories View Documents
Ricca Chemical Company
R710975020F
20 L Cubitainer
Each for $1,020.00
Add to Cart
 
Description

Description

Simulated Intestinal Fluid (without Pancreatin), CAS Number-7778-77-0, 1310-73-2, 500mL, 100.0, 0.66, 0.29, 97.0, 0.64, 0.27, Colorless, USP XXII Formulation, Poly natural, Liquid

Specifications

Specifications

7732-18-5, 7778-77-0, 1310-73-2
Laboratory
Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.