Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedical™ SimulTRAC-S Vitamin B12/Folate RIA Kit

Catalog No. MP06B254932 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
200 Tubes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP06B254932 200 Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP06B254932 Supplier MP Biomedical™ Supplier No. 06B254932

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Quantitative Solid Phase Radioimmunoassay

The SimulTRAC-S Vitamin B12/Folate RIA Kit is for the simultaneous quantitative determination of Vitamin B12 and Folate in serum.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Kit and Assay
Sample Type Plasma, Serum
Target Species Human
For Use With (Application) Endocrinology
Quantity 200 Tubes
Regulatory Status FDA Cleared (K822728/A)
Sample Volume 200 μL
Storage Requirements 2°C to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.