Learn More
Sin1 Polyclonal Antibody, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Chicken, Sheep, Bovine and Orangutan
Specifications
Specifications
| Antigen | Sin1 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.1% BSA and 0.09% sodium azide |
| Gene | MAPKAP1 |
| Gene Accession No. | Q9BPZ7 |
| Gene Alias | JC310, MEKK2-interacting protein 1, MGC2745, MIP1SIN1SIN1b, mitogen-activated protein kinase associated protein 1, Mitogen-activated protein kinase 2-associated protein 1, mSIN1, ras inhibitor MGC2745, SAPK-interacting protein 1, SIN1g, stress-activated protein kinase-interacting 1, stress-activated map kinase interacting protein 1, Stress-activated map kinase-interacting protein 1, target of rapamycin complex 2 subunit MAPKAP1, TORC2 subunit MAPKAP1 |
| Gene Symbols | MAPKAP1 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.