Learn More
Electron Microscopy Sciences Bench Top Single Bin Pipette Work Station
SDP
Supplier:  Electron Microscopy Sciences 6438509
Encompass_Preferred
Pipette work station withclear, hinged acrylic lid, clear acrylic bins for easy viewing, and self-closing lid. Holds 4 pipettors. Clear acrylic lids and fronts permit see-through storage. Work station may also be used to store small vials, caps, histo-cassettes, aluminum stubs. White. Measures: 9 x 9.5 x 6.75".
Catalog No. 50-305-79
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.