Learn More
SLINGSHOT BIOSCIENCES CD4(+), CD4(-) Single Biomarker Control
SDP
Supplier:  SLINGSHOT BIOSCIENCES SL00003
Encompass
Slingshot Labs single biomarker CD4(+), CD4(-) are lyophilized cell mimics that feature CD4+ biomarkers with scatter properties similar to lymphocyte populations.
Catalog No. NC2741754
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.