Learn More
INNOVATIVE BIOCHIPS 1CellAssay Single-Cell Isolation & Lysis Kit
Supplier:  INNOVATIVE BIOCHIPS K1SCLSKIT
Encompass_Preferred
1CellAssay Single-Cell Isolation & Lysis Kit provides a complete solution from easy single-cell isolation to rapid single-cell lysis in a small volume to perform various single-cell analyses, e.g. PCR, sequencing and multi-omics.Highlight: 1) Guarantee to isolate 24 single cells; 2) Isolate single cells in 2 µL volume; 3) Lyse single cells into 4 µL volumeFeatures: 1) Easy operation by a normal pipette in a sterile hood; 2) Gentle flow keeps high single-cell viability and integrity; 3) Compatible with cell size ≤ 50 µm and numbers ≤ 100 ; 4) Isolate and lyse 24 single cells in less than 30 minutes; 5) No special equipment or operation skills are required; 6) No liquid backflow or cross-talk between wells. Package list: 1CellDishTM-60mm, 2 devices; Cell Lysis Buffer, 200 µL; 8-strip PCR Tubes, 3 strips.
Catalog No. NC2131160
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.