Learn More
Perkin Elmer US LLC SMARTintro Organics Sample Introduction Kit - Orange
Supplier: Perkin Elmer US LLC N8150031
PerkinElmer introduces the NexION 2000 ICP-MS organics (Orange) solution-ready sample introduction technology. Engineered for the analysis of solvents, this organics optimized system incorporates quartz sample introduction components, organics torch, narrow injector, and addition gas line to enhance the analysis of samples containing organic materials. This kit includes: Quartz Concentric Type CT Nebulizer (N8152372) Cyclonic Quartz C3 High Sensitivity Spray Chamber with Matrix Gas Port (N8152383) One-Piece Organics Quartz Torch with 1.5 mm I.D. Injector (N8152616) Sample Line for MEINHARD Nebulizer (N8145503) Torch Light Shield Disk (N8152377) 0.5 mm I.D. MEINHARD Internal Standard Nebulizer Tee (N8152386) Cyclonic Spray Chamber/Cassette UV Light Shield (N8152425) Organics Addition Gas Line (N8152426) Orange Torch Cassette (N8152435) Torch Injector Nut Removal Tool (N8152443) Torch Injector Nut (N8152451) Internal Standard Probe (N8152452) PeriPump Union Male Fitting (N8152453) 0.8
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.