Learn More
Enzyme Research Laboratories Inc SINGLE CHAIN HIGH MOLECULAR
Supplier:  Enzyme Research Laboratories Inc HK 1300
Encompass
HK links prekallikrein to a negatively charged surface thereby allowing activation to kallikrein by surface bound Factor alpha -Xlla. The protein purity is determined by SDS-PAGE.
Catalog No. NC9937650
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.