Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ Single-Channel 96-well Temperature Verification Kit for PCR 9700 and 2720 Systems, 0.2 mL

Catalog No. 50591604
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-591-604 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-591-604 Supplier Applied Biosystems™ Supplier No. 4317939

This product is not available for sale online. Please contact Customer Service at 1-800-766-7000 or visit www.fishersci.com/quantstudio for more information and to contact a Life Science Specialist.

missing translation for 'validMainframeMsg'

Single-Channel 96-well Temperature Verification Kit for PCR 9700 and 2720 Systems, 0.2 mL

Temperature Verification Kits and Probes are engineered and manufactured specifically for Thermo Fisher Scientific to validate temperature calibration and uniformity in GeneAmp PCR systems. This kit is for use with the 96-Well GeneAmp PCR System 9700 and the GeneAmp PCR System 2720.
• Self-contained digital thermometer and battery operated for ease of use
• A platinum resistant probe (interchangable)

Complete offering
To expand the use of an existing Temperature Verification Kits, individual platinum resistant probes are also available in 0.02 ml, 0.1 ml, and 0.5 ml, for purchase separately.

More information can also be found at Alpha Technics (Formerly Alpha Sensors, Inc., Eutechnics Division). Calibration standards are traceable to NIST (US National Institute of Standards and Technology).

TRUSTED_SUSTAINABILITY

Specifications

Format Kit
Content And Storage This Temperature Verification System contains a Model 4500 digital thermometer (9V battery included), a 0.2 ml platinum resistant probe assembly, temperature verification frames to prevent ambient air from entering the sample block, light mineral oil, and cotton swabs. A carrying case is included.
For Use With (Equipment) 2720 Thermal Cycler, GeneAmp 9700
Product Type Temperature Verification Kit
For Use With (Application) PCR Amplification
Shipping Condition Room Temperature
Quantity 1 kit
Do you offer a temperature verification service to verify my PCR/qPCR instrument performance?

Yes, we offer an on-site temperature verification service with additional features such as ramp rate testing. Please find more information regarding the service below:
https://www.thermofisher.com/us/en/home/products-and-services/services/instrument-qualification-services/compliance-and-validation/on-site-temperature-verification.html

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.