Description
- Single Channel Protector For A Phone Line Or Small 14 Through 22 Gauge Wire
- Can Be Slit If Necessary (Supplied Unslit)
- Reduces Tripping Hazards
- Protects And Organizes Portable Cords And Cables
- Non-Metallic, Industrial-Grade Rubber Construction
- Multiple Sizes For Various Cable Diameters
- Grooved Underside For Better Traction
- Optional Connector Kit Available For Most Sizes
Specifications
Specifications
| Product Type | Cable Protector |
| Color | Black |
| Material | Rubber |