Learn More
Epredia™ Single Cytofunnel™ with White Filter Cards
Catalog No. 1102548 Shop All Epredia Products
Change view
Click to view available options
Quantity:
50 Pk.
500/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-025-48 500/Cs.
59-910-40 50 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 11-025-48 Supplier Epredia™ Supplier No. 1102548
Add to Cart
Add to Cart
This item is not returnable. View return policy
Hold sample volumes of up to 0.5 mL with the Epredia™ Single Cytofunnel™ with White Filter Cards.
  • Disposable sample chambers with white filter cards and caps
  • Pre-attached filter card
Specifications
Color White
Disposable Single-use
Quantity 500/Cs.
Type Single Cytofunnel™, With Filter Cards and Caps
Volume (Metric) 0.1 to 0.5 mL
WARNING: Cancer - www.P65Warnings.ca.gov