Learn More
Eisco™ Single Glass Cone
SCI_ED
Catalog No. S89372 Shop All Eisco Products
Change view
Click to view available options
Standard Taper Joint No.:
12/21
19/26
24/29
34/35
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No.
S89372 34/35
S89367 12/21
S89369 19/26
S89370 24/29
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S89372 Supplier Eisco™ Supplier No. CH0825G
Add to Cart
Add to Cart
A variety of standard cone sizes are available, each with a minimum shank length of 100 mm
  • Single cones used to connect glassware
  • Made from high quality borosilicate 3.3 glass
  • Cone features ground glass
  • Autoclavable
  • Warranty and Services
    1 year limited manufacturer warranty
    Specifications
    Cone Size 34/35
    For Use With (Application) Connect glassware
    Height (Metric) 10.6 cm
    Length (Metric) 3.5 cm
    Material Borosilicate Glass (3.3)
    Product Type Single Cone
    Quantity 1
    Standard Taper Joint No. 34/35