Description
- One layer of blue SMS
- Cross-stitch bond pattern
- Clear, visual feedback of breaches that may occur during handling, storage and/or sterilization
Specifications
Specifications
| Basis Weight | CH100 |
| Color | Blue |
| Material | SMS Fabric |
| Quantity | 500/Cs. |
| Dimensions (L x W) | 24 x 24 in. |
| Product Type | Medical Sterilization Wrap |