Learn More
Scinomix Inc SINGLE RACK 2ML CRYOVIAL
SDP
Supplier:  Scinomix Inc 1044500013SCI00010350
Encompass
Single Rack, 2ml CryovialRack is Landscape, 1x1 and fits VX2 or VXL. 6 columns and 8 rows. It is meant to work with 2mL Sarstedt tubes. It may be compatible with tubes of like size.
Catalog No. NC3593654
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.