Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Single Sided Meter Sticks
SCI_ED

Catalog No. p-7146831 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10 Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S88345 1 Ea.
S88346 10 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S88345 Supplier Eisco™ Supplier No. PH0064C
Only null left
Add to Cart
Add to Cart

Single Sided Meter Sticks are constructed from durable hardwood and feature a zero top and vertical reading.

  • Measurements graduated in centimeters and millimeters.
  • Dimensions: 100 cm x 2.5 cm x 6 mm
  • Ideal for the classroom.

Warranty and Services

1 year limited manufacturer warranty.

Specifications

Grade K to Undergraduate
For Use With (Application) Measuring
Includes 1 Meter Stick
Material Hardwood
Quantity 1 Ea.
Weight (English) 1.22 oz.
Length (English) 3.3 ft.
Length (Metric) 100 cm
Scale Metric
Class Size 1 Lab Group
Product Type Meter Scale
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.