Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propper Seracult™ Single Slide Fecal Occult Blood Test
SDP

Catalog No. 1255424B Shop All Propper Manufacturing Co. Products
Change view
Click to view available options
Quantity:
100 Pk.
1000 Pk.
Type:
Seracult Single Slide
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
12-554-24B 1000 Pk. Seracult Single Slide
12-554-24A 100 Pk. Seracult Single Slide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
For in vitro diagnostic use.
Catalog No. 12-554-24B Supplier Propper Supplier No. 37100200

missing translation for 'validMainframeMsg'

For in vitro diagnostic use.

Seracult™ Fecal Occult Blood Test is a qualitative aid in the diagnosis of colorectal cancer. Guaiac Fecal Occult Blood Test to aid in the detection of colorectal cancer. Single Slide is excellent for testing one sample. Long Sample Stability. Fast qualitative results.

  • Seracult™ Fecal Occult Blood Test is a qualitative aid in the diagnosis of colorectal cancer.
  • Detects 90% of colorectal cancers that bleed.
  • Designed for routine use in physical examinations in doctors' offices, hospitals, clinics, and laboratories or in the privacy and convenience of a patient's home.
  • Seracult™ Single Slides are ideal for in-office or inpatient is available in a variety of configurations to meet all of your needs.
  • Results are ready instantly.
  • Seracult paper allows for enhanced readability for early detection of even the smallest amount of blood in the sample.
  • Detects hemoglobin and products of its degradation.
  • Mailing kits of in-office slides.
  • 28-day sample stability.

Specifications

Clia Complexity Waived
Detectable Analytes Hemoglobin
Sample Type Stool
Type Seracult Single Slide
For Use With (Application) Colorectal Cancer Detection
Includes 1000 Single Slides, 20 x 15 mL Bottles of Developer, 1000 Applicators
Quantity 1000 Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.