Learn More
Description
- Material: Low-density polyethylene (LDPE)
- Features one thumb tab for easy removal of the cap
- Allows more tubes to be stacked closely in racks
- Maintain tight seal during centrifugation, agitation and refrigeration procedures
- Available in bright attractive colors for sample identification purposes
Specifications
Specifications
| Product Type | Snap Cap |
| Material | Polyethylene |
| Color | Yellow |
| No. per Pack | 1000 |
| For Use With (Equipment) | 10 mm blood drawing tubes, glass test tubes and plastic culture tubes |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.