Learn More
Thermco Products Single-use Humidity and Temperature Logger
Catalog No. 04500324
Change view
Click to view available options
Display Type:
Status Indicator Lights
Catalog No. Display Type
04-500-324 Status Indicator Lights
1 options
Catalog No. 04-500-324 Supplier Thermco Products Supplier No. LTHASO8
Add to Cart
Add to Cart
This single-use logger measures humidity from 0 to 100% Rh and temperature from -40°C to +85°C (-40°F to +185°F).
  • Housed in a durable polycarbonate case with a fast-reacting temperature sensor and an integrated real-time clock for date and time-stamped data.
  • Easily configured using the LogTag interface and free LogTag Analyzer software.
  • Supports delayed start, custom sampling intervals, and configurable alert conditions for the 'alert' indicator.
  • Recorded data can be charted, zoomed, analyzed, and exported to Excel.
  • Compliant with FDA 21 CFR Part 11, EC Annex 11, IATA 2018 Dangerous Goods Regulations, FCC, CE, RCM, and RoHS requirements.
  • The HASO-8 ensures accurate and reliable performance for temperature monitoring applications.
Specifications
Type Temperature and Humidity Datalogger
Display Type Status Indicator Lights
Alarm Types LED Indicator