Learn More
BURKLE INC Single-use sampler liquids
Supplier:  BURKLE INC 53931130
Encompass_Preferred
The LiquiDispo disposable sampler can be used to take samples of liquids from highly liquid to slightly viscous An ergonomic handle makes it easier to draw the sample up to 200 ml (6 76 oz ) can be drawn as with a syringe
Catalog No. 50-308-1302
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.