Learn More
Thermco Products Single-use USB Data Logger
Catalog No. 04500338
Change view
Click to view available options
Alarm Types:
LED Indicator
Catalog No. Alarm Types
04-500-338 LED Indicator
1 options
Catalog No. 04-500-338 Supplier Thermco Products Supplier No. SRIC4
Add to Cart
Add to Cart
Single-use USB Data Logger records a temperature range between -30 °C to +60 °C (-22 °F to +140 °F).
  • The LogTag™ SRIC-4 Single-Trip Temperature Data Logger is designed for monitoring temperature-sensitive products during short or long-haul transportation.
  • Measures temperatures from -30°C to 60°C with an accuracy of ±0.5°C between -5°C and 30°C, and a resolution of 0.1°C.
  • Stores up to 3,968 temperature readings, supporting approximately 13 days at 5 min. intervals or 41 days at 15 min. intervals.
  • Powered by a CR2032 battery with an operating life of up to 20 Months.
  • Configurable using LogTag Analyzer software and a standard LogTag Interface Cradle.
  • Supports flexible sampling intervals from 1 min. to 18 hr. and alarm settings with one upper and one lower limit.
  • Flashing OK and ALERT indicators provide instant visual status checks.
  • Housed in a durable IP65-rated polycarbonate case measuring 77 x 54.5 x 8.6 mm and weighing 28 g.
  • Integrated real-time clock ensures accurate time-stamped data.
  • Ideal for food, pharmaceutical, and logistics sectors, with support for data charting, archiving, and export for effective temperature management.
Specifications
Type Single-use Datalogger
Display Type No Display
Alarm Types LED Indicator