Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Single Well High Profile Reagent Reservoirs
GREENER_CHOICE

Catalog No. 14222411 Shop All Axygen Products
Change view
Click to view available options
Capacity (Metric):
240 mL
282 mL
290 mL
Sterility:
Non-sterile
Sterile
Channels:
8
12
96
384
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Sterility Channels
14-222-411 290 mL Non-sterile 12
14-222-418 282 mL Sterile 384
14-222-422 290 mL Sterile 8
14-222-412 290 mL Sterile 12
14-222-421 290 mL Non-sterile 8
14-222-423 240 mL Non-sterile 96
14-222-424 240 mL Sterile 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 14-222-411 Supplier Axygen™ Supplier No. RESSW12HP
Add to Cart
Add to Cart

Manufactured with virgin polypropylene for excellent chemical resistance

Corning offers a complete line of Axygen single-well and multiple-well reagent reservoirs which are available in multiple formats for automated and manual use. All reservoirs are designed to comply with standard ANSI/SLAS footprint dimensions for microplates and are compatible with most automation workstations. Reagent reservoirs are manufactured with polypropylene for excellent chemical resistance.

  • Durable and cost effective
  • Various formats for multiple applications
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ANSI/SLAS, RNase/DNase-free, non-pyrogenic
Color Clear
Capacity (Metric) 290 mL
Material Polypropylene
Sterility Non-sterile
Type Reagent Reservoir
No. per Case 25/Cs.
No. per Pack 25/Pk.
Packaging 25/Cs.
Channels 12
Disposable Single-use
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.