Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Single Well Low Profile Reagent Reservoirs
GSA_VAGREENER_CHOICE

Catalog No. 14222419 Shop All Axygen Products
Change view
Click to view available options
Capacity (Metric):
86 mL
90 mL
92 mL
Sterility:
Non-sterile
Sterile
Channels:
1
96
384
Packaging:
25/Cs.
Individually Wrapped; 25/Cs.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Sterility Channels Packaging
14-222-419 92 mL Non-sterile 384 25/Cs.
14-222-425 86 mL Non-sterile 96 25/Cs.
14-222-426 86 mL Sterile 96 Individually Wrapped; 25/Cs.
14-222-413 90 mL Non-sterile 1 25/Cs.
14-222-420 92 mL Sterile 384 Individually Wrapped; 25/Cs.
14-222-414 90 mL Sterile 1 Individually Wrapped; 25/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 14-222-419 Supplier Axygen™ Supplier No. RESSW384LP
Add to Cart
Add to Cart

Manufactured with virgin polypropylene for excellent chemical resistance

Corning offers a complete line of Axygen single-well and multiple-well reagent reservoirs which are available in multiple formats for automated and manual use. All reservoirs are designed to comply with standard ANSI/SLAS footprint dimensions for microplates and are compatible with most automation workstations. Reagent reservoirs are manufactured with polypropylene for excellent chemical resistance.

  • Durable and cost effective
  • Various formats for multiple applications

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ANSI/SLAS, RNase/DNase-free, non-pyrogenic
Color Clear
Capacity (Metric) 92 mL
Material Polypropylene
Sterility Non-sterile
Type Reagent Reservoir
Packaging 25/Cs.
Channels 384
Disposable Single-use
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.