Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Sir2.1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01950247
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-950-247 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-950-247 Supplier Invitrogen™ Supplier No. PA116933
  • $626.00
    $506.90 / Each of 1

    Save $119.10

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sir2.1 Polyclonal Antibody for Western Blot, IHC (P), IP

Sir2 (silent information regulator 2) protein plays a crucial role in regulating the life span of C. elegans by means of caloric restricion. Overexpression of the protein can extend the life span, and is, thus, a regulator of longevity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sir2.1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum with 0.05% sodium azide
Gene sir-2.1
Gene Accession No. Q21921
Gene Alias CELE_R11A8.4; Deacetylase sirtuin-type domain-containing protein;NAD-dependent protein deacetylase sir-2.1; NAD-dependent deacetylase sirtuin-2; NAD-dependent protein deacetylase sir-2.1; NAD-dependent protein deacetylase sirtuin-2; Protein sir-2.1; protein-coding; R11A8.4; Regulatory protein SIR2 homolog 1; Regulatory protein SIR2 homolog 2; silent information regulator 2; SIR2; sir-2.1; SIR2L; SIR2L2; SIR2-like protein 2; sir2-related protein type 2; SIRT2; sirtuin 2; sirtuin type 2; sirtuin-2
Gene Symbols sir-2.1
Host Species Rabbit
Immunogen A full-length recombinant protein to Sir2.1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 177924
Target Species C. elegans
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.