Learn More
BioVendor, LLC Sircol; Dye Reagent (3 x 110 ml)
SDP
Supplier:  BioVendor, LLC S1005
Encompass
Sircol; Dye Reagent (3 x 110 ml). Contains Sirius Red in picric acid and has been formulated for specific binding to collagen under the conditions defined in the Sircol Manual. Sircol™ Collagen Assay component. Biocolor Item # S1005
Catalog No. 50-172-5955