Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIRP beta/gamma Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA5148568
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA5148568 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA5148568 Supplier Invitrogen™ Supplier No. PA5148568
  • $499.00
    $374.65 / Each of 1

    Save $124.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

SIRP beta and SIRP gamma are two members of the signal regulatory protein (SIRP) family. These proteins are transmembrane receptors that are expressed on the surface of various cells, including immune cells and neurons. The function of SIRP beta and SIRP gamma is to regulate cellular signaling pathways that are involved in immune responses, cell adhesion, and phagocytosis. Specifically, these proteins interact with the protein tyrosine phosphatase SHP-1, which is a negative regulator of immune cell activation. By binding to SHP-1, SIRP beta and SIRP gamma can inhibit immune cell activation and promote immune tolerance. In addition to their role in immune regulation, SIRP beta and SIRP gamma have also been implicated in neuronal signaling and synaptic plasticity. These proteins are thought to play a role in the formation and maintenance of synapses, as well as in the regulation of neuronal activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIRP beta/gamma
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with trehalose and no preservative
Gene SIRPB1
Gene Accession No. O00241, Q9P1W8
Gene Symbols SIRPB1, SIRPG
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human SIRP beta 1, Gly26-Ala369 (Arg53His and Ala363Pro).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10326, 55423
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.