Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIRP gamma Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581025 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581025 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581025 Supplier Invitrogen™ Supplier No. PA581025
  • $411.50
    $331.65 / Each of 1

    Save $79.85

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human SIRPG/SIRP gamma/CD172g.

SIRP gamma (CD172g) recognizes a 55 kDa signal regulatory protein which is a member of the immunoglobulin gene superfamily. SIRP gamma is expressed on most T lymphocytes and a subset of B-cells but is absent on myeloid cells. CD172g has a truncated cytoplasmic tail, which is similar to SIRP beta, but unlike SIRP beta CD172g does not require DAP12 for expression at the cell surface. CD172g, like SIRP alpha, binds to CD47 inducing apoptosis. SIRP gamma is also involved in the negative regulation of receptor tyrosine kinase-coupled signalling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIRP gamma
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 5.35 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene SIRPG
Gene Accession No. Q9P1W8
Gene Alias bA77C3.1; CD172 antigen-like family member B; CD172 gamma; CD172g; CD172g antigen; RP11-77C3.2; signal regulatory protein gamma; signal-regulatory protein beta 2; signal-regulatory protein beta-2; Signal-regulatory protein gamma; SIRP beta 2; SIRPB2; SIRP-B2; SIRP-beta-2; SIRPG; SIRPgamma; SIRP-gamma
Gene Symbols SIRPG
Host Species Rabbit
Immunogen Recombinant Human SIRPG/SIRP gamma/CD172g protein (Met1-Ser364).
Purification Method Protein A, Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55423
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.