Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIRT1 Monoclonal Antibody (OTI12H3), TrueMAB™, OriGene
SDP

Catalog No. 501683060
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-3060 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-3060 Supplier OriGene Supplier No. TA809859
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NAD-dependent protein deacetylase sirtuin-1 (SIRT1) links transcriptional regulation directly to intracellular energetics and participates in the coordination of several separated cellular functions such as cell cycle, repsonse to DNA damage, metobolism, apoptosis, and authopahy. SIRT1 can modulate chromatin function through deacetylation of histones and can promote alterations in the methylation of histone and DNA, leading to transcriptional repression. It is essential in skeletal muscle cell differentiation and in response to low nutrients mediates the inhibitory effect on skeletal myoblast differentiation which also involves 5′-AMP-activated protein kinase (AMPK) and nicotinamide phosphoribosyltransferase (NAMPT).

Specifications

Antigen SIRT1
Applications Western Blot
Classification Monoclonal
Clone OTI12H3
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SIRT1
Gene Accession No. Q96EB6
Gene Alias SIR2, SIR2L1, SIR2alpha
Gene Symbols SIRT1
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 552-676 of human SIRT1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23411
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.