Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sirt1 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501558221
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-8221 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-8221 Supplier Bethyl Laboratories, Inc Supplier No. A300688AT
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

SIRT1, the human homolog of the S. cerevisiae Sir2 protein, functions as an NAD-dependent deacetylase of a number of nonhistone substrates including p53. In response to DNA damage, SIRT1 binds and deacetylates the p53 protein at c-terminal Lys382 residue and attenuate p53-mediated functions. When overexpressed in mouse embryo fibroblasts, SIRT1 antagonizes PML-induced acetylation of p53 and rescues PML-mediated premature cellular senescence. In mammalian cells, SIRT1 appears to control the cellular response to stress by regulating the FOXO family of forkhead transcription factors.

Specifications

Antigen Sirt1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene SIRT1
Gene Accession No. Q96EB6
Gene Alias NAD-dependent deacetylase sirtuin-1; OTTHUMP00000198111; OTTHUMP00000198112; SIR2-like protein 1; SIR2alpha; hSIR2; hSIRT1; sir2-like 1; sirtuin type 1
Gene Symbols SIRT1
Host Species Rabbit
Immunogen Between 700 and C-term
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23411
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.