Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam SIRT3 Activity Assay Kit, Fluorometric, Human, 100 Tests

Catalog No. 502582856
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-2856 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-2856 Supplier Abcam Supplier No. AB156067100TEST
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rapidly measure SIRT3 activity in cell and tissue extracts within 40 minutes.

  • Assay Type: Enzyme activity
  • Detection Method: Fluorescent
  • Sample Types: Purified protein, Mitochondrial Extract
  • Species Reactivity: Human
  • Assay Time: 40 min
  • Applications: Enzyme activity assay

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Run Time 40 min.
Reactivity Human
Target Molecule SIRT3
No. of Reactions 100 Tests
For Use With (Equipment) Fluorometric Plate Reader
Format Kit
Detection Location In-Solution Detection
Detection Method Fluorescence
Cross Reactivity Human
Sample Type Purified Protein, Mitochondrial Extract
Quantity 100 Tests
For Use With (Application) Enzyme activity assay
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.