Learn More
Abcam SIRT6 Activity Assay Kit, Fluorometric, Human, 100/Test
Description
- Assay Type: Enzyme activity
- Detection Method: Fluorescent
- Assay Time: 1 hr
- Reactive Species: Human
- Sample Types: Recombinant protein, purified protein
- Applications: Enzyme activity assay
- Citations: Cited in > 8 publications
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | SIRT6 |
| Reactivity | Human |
| No. of Reactions | 100 Tests |
| For Use With (Equipment) | Microplate Reader |
| Format | Kit |
| Detection Method | Fluorescence |
| Detection Location | In-Solution Detection |
| Cross Reactivity | Human |
| Sample Type | Recombinant Protein, Purified protein |
| Quantity | 100 Tests |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.