Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIRT6 Monoclonal Antibody (2G1H1)
GREENER_CHOICE

Catalog No. PIMA1214 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA1214 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA1214 Supplier Invitrogen™ Supplier No. MA1214
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

MA1-214 SIRT6 antibody detects SIRT6 by western blot and immunofluroescence. MA1-214 SIRT6 antibody detects SIRT6 by western blot at ~42kDa.

Sirtuins regulate epigenetic gene silencing. SIRT6 (Mono-ADP-ribosyltransferase sirtuin-6), a mouse Sir2 homolog, is a chromatin-associated nuclear ADP-ribosyltransferase protein. It appears to be involved in DNA repair and may also play a role in human aging.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIRT6
Applications Western Blot, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 2G1H1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene SIRT6
Gene Accession No. Q8N6T7
Gene Alias 2810449N18Rik; AI043036; NAD-dependent deacetylase sirtuin-6; NAD-dependent protein deacetylase sirtuin-6; Regulatory protein SIR2 homolog 6; SIR2; SIR2 like 6; Sir2l6; SIR2-like protein 6; sir2-related protein type 6; Sirt6; sirtuin 6; sirtuin type 6
Gene Symbols SIRT6
Host Species Mouse
Immunogen Purified recombinant fragment of human SIRT6 (AA: 141-250) expressed in E. Coli.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51548
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.