Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIRT6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01950186
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-950-186 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-950-186 Supplier Invitrogen™ Supplier No. PA116957
  • $587.00
    $487.70 / Each of 1

    Save $99.30

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-16957 is specific for the ~39 kDa SIRT6 protein. A higher molecular weight band at ~59 kDa may be seen with lower dilutions such as 2 μg/mL. Suggested positive control: antigen standard for SIRT6 (transient overexpression lysate), Hela whole cell extract.

Sirtuins regulate epigenetic gene silencing. SIRT6 (Mono-ADP-ribosyltransferase sirtuin-6), a mouse Sir2 homolog, is a chromatin-associated nuclear ADP-ribosyltransferase protein. It appears to be involved in DNA repair and may also play a role in human aging.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIRT6
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.05 mg/mL
Conjugate Unconjugated
Formulation Phosphate, tris citrate with 0.1% sodium azide; pH 7-8
Gene SIRT6
Gene Accession No. Q8N6T7
Gene Alias 2810449N18Rik; AI043036; NAD-dependent deacetylase sirtuin-6; NAD-dependent protein deacetylase sirtuin-6; Regulatory protein SIR2 homolog 6; SIR2; SIR2 like 6; Sir2l6; SIR2-like protein 6; sir2-related protein type 6; Sirt6; sirtuin 6; sirtuin type 6
Gene Symbols SIRT6
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region of the human SIRT6 protein (within residues 300-355).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51548
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.