Learn More
Selleck Chemical LLC Sirtinol 5mg 410536-97-9
SDP
Supplier:  Selleck Chemical LLC S28045MG
Encompass_Preferred
Sirtinol is a specific SIRT1 and SIRT2 inhibitor with IC50 of 131 ?M and 38 ?M in cell-free assays, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0877023
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.