Learn More
AdipoGen Sirtuin 5 human pAb
SDP
Supplier:  AdipoGen AG25A0100C100
Encompass_Preferred
Polyclonal Antibody. Recognizes human sirtuin 5 intact form. Detects a band of ~37kDa by Western blot. Source Rabbit. Applications ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. Sirtuin 5 is a NAD-dependent protein deacetylase that activates CPS1 and contributes to the regulation of blood ammonia levels during fasting. Can deacetylate cytochrome c and a number of other proteins in vitro.
Catalog No. 50-256-3220
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.