Learn More
AdipoGen Sirtuin 6 human pAb
SDP
Supplier:  AdipoGen AG25A0101C100
Encompass_Preferred
Polyclonal Antibody. Recognizes human sirtuin 6. Detects a band of ~42kDa by Western blot. Source Rabbit. Applications ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. Sirtuin 6 SIRT6 is a NAD-dependent protein deacetylase with activity towards Lys-9 and Lys-56 of histone H3. It modulates acetylation of histone H3 in telomeric chromatin during the S-phase of the cell cycle. It is required for genomic stability and for normal IGF1 serum levels and normal glucose homeostasis. It also regulates cellular senescence and apoptosis. Unlike most sirtuins, SIRT6 preferentially removes long-chain fatty-acyl, rather than acetyl, protein modifications. This activity regulates secretion of the inflammation-associated protein TNF-alpha.
Catalog No. 50-256-3219
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.