Learn More
Medchemexpress LLC HY-128918 5mg Medchemexpress, SIS17 CAS:2374313-54-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1289185MG
Encompass_Preferred
Medchemexpress, HY-128918 5mg SIS17 CAS:2374313-54-7 SIS17 is a mammalian histone deacetylase 11 (HDAC 11) inhibitor with an IC50 value of 0.83 μM, inhibits the demyristoylation HDAC11 substrate, serine hydroxymethyl transferase 2, without inhibiting other HDACs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9783
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.