Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIT Monoclonal Antibody (SIT-01), PE
GREENER_CHOICE

Catalog No. 01676379 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-676-379 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-676-379 Supplier Invitrogen™ Supplier No. MA119652
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

This antibody reacts with murine SIT.

SIT (SHP2-interacting transmembrane adaptor protein) is expressed exclusively in lymphoid organs and acts either as a positive or as a negative regulatory element in T cell activation and in T cell development. Binding to Grb2 plays a pivotal role in signal transduction. Hubener et al. (2001) determined that the SIT gene contains 5 exons and spans 1.8 kb of genomic DNA. The SIT promoter demonstrated strong transcriptional activity and potential binding sites for both ubiquitous and lymphoid-specific transcription factors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIT
Applications Flow Cytometry
Classification Monoclonal
Clone SIT-01
Concentration 0.1 mg/mL
Conjugate PE
Formulation PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
Gene SIT1
Gene Accession No. Q9Y3P8
Gene Alias gp30/40; MGC125908; MGC125909; MGC125910; RP11-331F9.5; SHP2 interacting transmembrane adaptor; SHP-2 interacting transmembrane adaptor protein; SHP2-interacting transmembrane adapter protein; SHP2-interacting transmembrane adaptor protein; signaling threshold regulating transmembrane adaptor 1; signaling threshold-regulating transmembrane adapter 1; SIT; SIT1; SIT-R; suppression inducing transmembrane adaptor 1; suppression-inducing transmembrane adapter 1
Gene Symbols SIT1
Host Species Mouse
Immunogen Bacterially produced recombinant intracellular fragment of human SIT.
Purification Method Size-exclusion chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27240
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.