Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SiteClick™ Antibody Azido Modification Kit

Catalog No. S10900
Encompass_Preferred
Change view
Click to view available options
Labeling Scale:
1 mg
100 to 250 μg
5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Labeling Scale
S10900 1 mg
S10901 5 mg
S20026 100 to 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S10900 Supplier Invitrogen™ Supplier No. S10900
Add to Cart
Edge
Add to Cart

Create a label-ready site-specific azido-modified antibody without lengthy genetic modification using the SiteClick Antibody Azido Modification Kit.

Create a label-ready, site-specific, azido-modified antibody without lengthy genetic modification using the SiteClick Antibody Azido Modification Kit. Use this kit to carry out a two-step conjugation process over three days. SiteClick labeling uses enzymes to attach an azido moiety to the heavy chains of an IgG antibody, ensuring the antigen-binding domains remain unaltered.

Create a label-ready, site-specific, azido-modified antibody without lengthy and inefficient genetic modification using the SiteClick Antibody Azido Modification Kit. SiteClick labeling uses enzymes to specifically attach an azido moiety to the heavy chains of an IgG antibody, ensuring that the antigen-binding domains remain unaltered for binding to the antigen target. This site selectivity is achieved by targeting the carbohydrate domains present on the Fc region of essentially all IgG antibodies regardless of isotype and host species.

Once azido–modified, a variety of SiteClick sDIBO or DBCO alkyne labels are available for covalent attachment to the antibody via copper-free click chemistry under mild reaction conditions at your choice of scale. This provides the flexibility to choose different fluorescent labels for your antibody, attach another molecule via streptavidin, or attach your own molecule, such as an oligo, drug, or toxin via amine-reactive or amine-containing moieties depending on your assay. This site-specific modification using SiteClick labeling is a critical tool in the development of therapeutic antibodies, antibody-drug conjugates, biologics, and immunodiagnostic tests that require high specificity with reproducible labeling reactions and helps ensure that antibody performance and function are preserved after conjugation.

Features of the SiteClick Antibody Azido Modification Kit include:
• Contains everything required to azido-modify any species or isotype of IgG antibody
• Easy-to-follow step-by-step protocol
• Highly efficient, site-specific labeling preserves antibody function and results in high quality antibody conjugate
• Reproducible labeling chemistry with lot-to-lot consistency in degree of labeling
• Optimal method for site-specific labeling of therapeutic antibodies, antibody-drug conjugates, biologics, and immunodiagnostics

Custom SiteClick Antibody Labeling Service and sDIBO labels
If you have an antibody that is considered 'difficult to label' or has lost activity after labeling using a conventional method, please contact our custom service representatives to determine whether the SiteClick Antibody Labeling Service would be right for your antibody. We offer complete custom SiteClick antibody labeling services with the option of multiple detection molecules including biotin, Alexa Fluor dyes, Qdot fluorophores, R-PE, chelates for PET imaging, and many others.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Antibody Azido Modification Kit
Quantity for 1 mg of Ab
Content And Storage Store at 2°C to 8°C. Do not freeze.
Labeling Scale 1 mg
Shipping Condition Wet Ice
Product Line SiteClick
Chemical Reactivity Azide
Reactive Moiety Azide
Label or Dye Alexa Fluor 488, Alexa Fluor 647, Alexa Fluor 555
Color Clear
Labeling Target Antibodies
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.