Specifications
| Certifications/Compliance | ANSI Z87.1,CSA Z94.3 |
| Certifications/Compliance (Europe) | EN-166 |
| Lens Tint | Gray |
| Frame Color | Gray, Lime |
| Weight (Metric) | 24.4 g |
| Lens Material | Polycarbonate |
| Frame Material | Polycarbonate |